ENA379363573 Enamine

2-methyl-4-nitro-N-(pyrrolidin-3-yl)benzamide hydrochloride

  • MDL number MFCD22578656
  • Cas Number : 1423033-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118021

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118021
Availability details for
Quantity
  • Availability
  • Add to Cart