ENA379363575 Enamine

tert-butyl N-(4-sulfanylphenyl)carbamate

  • MDL number MFCD22578659
  • Cas Number : 852448-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118090

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118090
Availability details for
Quantity
  • Availability
  • Add to Cart