ENA379363578 Enamine

2-tert-butyl-1-(2-phenoxyethyl)piperazine dihydrochloride

  • MDL number MFCD22578663
  • Cas Number : 1423024-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118112

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118112
Availability details for
Quantity
  • Availability
  • Add to Cart