ENA379363579 Enamine

1-(5-bromo-2-fluorophenyl)propan-2-amine hydrochloride

  • MDL number MFCD22578664
  • Cas Number : 1423034-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118129
Availability details for
Quantity
  • Availability
  • Add to Cart