ENA379363587 Enamine

1,1,3,3-tetramethyl-2,3-dihydro-1H-inden-5-amine hydrochloride

  • MDL number MFCD22578672
  • Cas Number : 1423027-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118225
Availability details for
Quantity
  • Availability
  • Add to Cart