ENA379363590 Enamine

methyl 2-[(2-aminoethyl)(methyl)amino]acetate dihydrochloride

  • MDL number MFCD22578677
  • Cas Number : 1423028-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118296
Availability details for
Quantity
  • Availability
  • Add to Cart