ENA379363591 Enamine

2-[(2-methylpropyl)sulfanyl]ethan-1-amine hydrochloride

  • MDL number MFCD22578678
  • Cas Number : 1423028-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118297

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118297
Availability details for
Quantity
  • Availability
  • Add to Cart