ENA379363592 Enamine

3-amino-1-(morpholin-4-yl)-3-phenylpropan-1-one hydrochloride

  • MDL number MFCD22578680
  • Cas Number : 1423028-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118313

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118313
Availability details for
Quantity
  • Availability
  • Add to Cart