ENA379363595 Enamine

5-methyl-5-(pyridin-3-yl)imidazolidine-2,4-dione hydrochloride

  • MDL number MFCD22578683
  • Cas Number : 1423028-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118363

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118363
Availability details for
Quantity
  • Availability
  • Add to Cart