ENA379363599 Enamine

1-[2-(2-chlorophenoxy)ethyl]-1H-indole-2-carboxylic acid

  • MDL number MFCD22578688
  • Cas Number : 1423032-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118454

Safety Information

signalword Warning
pcodes P264,P270,P301+P312,P330,P501
MSDS https://www.enaminestore.com/catalog/EN300-118454
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart