ENA379363610 Enamine

1-(2,3,4,5-tetrahydro-1H-3-benzazepin-7-yl)ethan-1-one hydrochloride

  • MDL number MFCD22578702
  • Cas Number : 34685-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118518
Availability details for
Quantity
  • Availability
  • Add to Cart