ENA379363612 Enamine

1-ethyl-2-(methylsulfanyl)-1H-imidazo[4,5-c]pyridine

  • MDL number MFCD22578704
  • Cas Number : 1423032-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118520

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118520
Availability details for
Quantity
  • Availability
  • Add to Cart