ENA379363614 Enamine

[4-(methanesulfonylmethyl)oxan-4-yl]methanol

  • MDL number MFCD22578706
  • Cas Number : 1423034-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118542

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118542
Availability details for
Quantity
  • Availability
  • Add to Cart