ENA379363615 Enamine

3-(phenoxymethyl)-5-(trichloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD22578707
  • Cas Number : 1423031-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118543
Availability details for
Quantity
  • Availability
  • Add to Cart