ENA379363622 Enamine

N-(3-aminopropyl)-2-methylpropanamide hydrochloride

  • MDL number MFCD22578715
  • Cas Number : 1423032-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118656
Availability details for
Quantity
  • Availability
  • Add to Cart