ENA379363644 Enamine

4-methyl-7-phenoxy-1H,2H,3H,4H,9H-cyclopenta[b]quinolin-9-one

  • MDL number MFCD02115681
  • Cas Number : 374918-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116780

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116780
Availability details for
Quantity
  • Availability
  • Add to Cart