ENA379363656 Enamine

2-chloro-4-(chloromethyl)-1-methoxybenzene

  • MDL number MFCD00068624
  • Cas Number : 13719-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 38-39
Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116796
Availability details for
Quantity
  • Availability
  • Add to Cart