ENA379363658 Enamine

N-(2-carbamothioylethyl)-N-methylacetamide

  • MDL number MFCD11130020
  • Cas Number : 1019345-48-2
  • Molecular Weight : 160.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-116798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116798
Availability details for
Quantity
  • Availability
  • Add to Cart