ENA379363667 Enamine

2-(chloromethyl)-4-methylpyrimidine

  • MDL number MFCD10697342
  • Cas Number : 78060-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116810
Availability details for
Quantity
  • Availability
  • Add to Cart