ENA379363681 Enamine

[2-(1H-pyrazol-1-yl)phenyl]methanamine hydrochloride

  • MDL number MFCD13186034
  • Cas Number : 1269225-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-116840

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116840
Availability details for
Quantity
  • Availability
  • Add to Cart