ENA379363689 Enamine

2-bromobicyclo[2.2.1]heptane-1-carboxylic acid

  • Cas Number : 74830-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113677

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113677
Availability details for
Quantity
  • Availability
  • Add to Cart