ENA379363697 Enamine

1-(4-tert-butylphenyl)cyclopentan-1-amine

  • MDL number MFCD11899007
  • Cas Number : 160001-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116891

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116891
Availability details for
Quantity
  • Availability
  • Add to Cart