ENA379363710 Enamine

3-(morpholin-4-yl)-1,2,4-thiadiazol-5-amine

  • MDL number MFCD09865005
  • Cas Number : 138588-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-116914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116914
Availability details for
Quantity
  • Availability
  • Add to Cart