ENA379363712 Enamine

N'-hydroxy-3-(2-methylmorpholin-4-yl)-3-oxopropanimidamide

  • Cas Number : 1094299-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-116917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116917
Availability details for
Quantity
  • Availability
  • Add to Cart