ENA379363713 Enamine

2-(1-methylcyclohexyl)acetic acid

  • MDL number MFCD14529334
  • Cas Number : 14352-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-116918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116918
Availability details for
Quantity
  • Availability
  • Add to Cart