ENA379363729 Enamine

3,4-dihydrospiro[1-benzopyran-2,1'-cyclohexan]-4-one

  • MDL number MFCD18431738
  • Cas Number : 62756-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116961
Availability details for
Quantity
  • Availability
  • Add to Cart