ENA379363735 Enamine

1-(2-tert-butylphenyl)ethan-1-one

  • MDL number MFCD19369793
  • Cas Number : 22583-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116967

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116967
Availability details for
Quantity
  • Availability
  • Add to Cart