ENA379363740 Enamine

tert-butyl pyrazolidine-1-carboxylate

  • MDL number MFCD09817426
  • Cas Number : 57699-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114089

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114089
Availability details for
Quantity
  • Availability
  • Add to Cart