ENA379363756 Enamine

2-[3-(2-methylpropoxy)phenoxy]acetic acid

  • MDL number MFCD01409169
  • Cas Number : 392237-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114459

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114459
Availability details for
Quantity
  • Availability
  • Add to Cart