ENA379363773 Enamine

2-(1,2-oxazol-3-yl)ethan-1-amine hydrochloride

  • MDL number MFCD19160593
  • Cas Number : 1314902-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-130
Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117053
Availability details for
Quantity
  • Availability
  • Add to Cart