ENA379363783 Enamine

(1S)-1-(2-methyl-1,3-thiazol-4-yl)ethan-1-ol

  • MDL number MFCD18339613
  • Cas Number : 1344929-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114745
Availability details for
Quantity
  • Availability
  • Add to Cart