ENA379363793 Enamine

8-oxononanoic acid

  • MDL number MFCD20638209
  • Cas Number : 25542-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117102
Availability details for
Quantity
  • Availability
  • Add to Cart