ENA379363795 Enamine

4-chloro-2-(1H-pyrrole-2-carbonyl)aniline

  • MDL number MFCD00191729
  • Cas Number : 74978-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-117104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117104
Availability details for
Quantity
  • Availability
  • Add to Cart