ENA379363796 Enamine

6-methyl-1,4-oxazepane

  • MDL number MFCD18791235
  • Cas Number : 1246494-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117105
Availability details for
Quantity
  • Availability
  • Add to Cart