ENA379363801 Enamine

7-chloro-3-methyl-1lambda6-benzothiophene-1,1-dione

  • MDL number MFCD22578564
  • Cas Number : 17514-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117110
Availability details for
Quantity
  • Availability
  • Add to Cart