ENA379363817 Enamine

1-(3-methoxyphenyl)-2-methylpropan-2-amine

  • MDL number MFCD09888582
  • Cas Number : 198226-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117132
Availability details for
Quantity
  • Availability
  • Add to Cart