ENA379363839 Enamine

piperidin-4-yl acetate hydrochloride

  • MDL number MFCD08061428
  • Cas Number : 94886-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117170

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117170
Availability details for
Quantity
  • Availability
  • Add to Cart