ENA379363854 Enamine

2,6-dichloro-[1,3]oxazolo[4,5-b]pyridine

  • MDL number MFCD18262175
  • Cas Number : 1211529-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117200
Availability details for
Quantity
  • Availability
  • Add to Cart