ENA379363856 Enamine

1-bromo-3-[(propan-2-yloxy)methyl]benzene

  • MDL number MFCD14687023
  • Cas Number : 1247851-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117202
Availability details for
Quantity
  • Availability
  • Add to Cart