ENA379363863 Enamine

methyl 2-cyano-3-methylbenzoate

  • MDL number MFCD18399106
  • Cas Number : 500024-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117210

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117210
Availability details for
Quantity
  • Availability
  • Add to Cart