ENA379363870 Enamine

N-(2-cyanoethyl)-N-(propan-2-yl)acetamide

  • MDL number MFCD16053907
  • Cas Number : 4271-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117218
Availability details for
Quantity
  • Availability
  • Add to Cart