ENA379363871 Enamine

4-methyl-2-(1H-1,2,4-triazol-3-yl)aniline

  • MDL number MFCD14640957
  • Molecular Weight : 174.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-117219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117219
Availability details for
Quantity
  • Availability
  • Add to Cart