ENA379363877 Enamine

5-[hydroxy(piperidin-4-yl)methyl]-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD11186080
  • Cas Number : 1094442-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117226
Availability details for
Quantity
  • Availability
  • Add to Cart