ENA379363917 Enamine

4-(3,4-dihydro-2H-1-benzopyran-6-yl)-5-methyl-1,3-thiazol-2-amine

  • MDL number MFCD10006115
  • Cas Number : 1017398-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-117376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117376
Availability details for
Quantity
  • Availability
  • Add to Cart