ENA379363922 Enamine

2-(oxolan-3-yl)-1-phenylethan-1-one

  • MDL number MFCD21923241
  • Cas Number : 1423024-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117382

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117382
Availability details for
Quantity
  • Availability
  • Add to Cart