ENA379363923 Enamine

methyl 4-{[(3,5-dimethyl-1,2-oxazol-4-yl)methyl](methyl)amino}butanoate

  • MDL number MFCD15506907
  • Cas Number : 1423034-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117383

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117383
Availability details for
Quantity
  • Availability
  • Add to Cart