ENA379363925 Enamine

N-[(pyridin-3-yl)methyl]pyrrolidine-2-carboxamide

  • MDL number MFCD11120235
  • Cas Number : 1132659-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117385
Availability details for
Quantity
  • Availability
  • Add to Cart