ENA379363931 Enamine

(3R)-1-[(3-phenyl-1,2,4-oxadiazol-5-yl)methyl]pyrrolidin-3-ol

  • MDL number MFCD22578596
  • Cas Number : 1423040-64-5
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117398
Availability details for
Quantity
  • Availability
  • Add to Cart