ENA379363939 Enamine

4-phenyl-1H-indole

  • MDL number MFCD13177532
  • Cas Number : 35577-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115897

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-115897
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart