ENA379363940 Enamine

methyl 3-amino-4-(4-methyl-1H-pyrazol-1-yl)benzoate

  • MDL number MFCD13343553
  • Cas Number : 1291529-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117416
Availability details for
Quantity
  • Availability
  • Add to Cart